Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-O-Palmitoyl-2-O-acetyl-sn-glycero-3-phosphocholine, 99%

Click to view available options
Quantity:
1mg
5mg
84062-61-3
C26H52NO8P
537.675
MFCD00798242
XPAXRSJGGFVTFM-VWLOTQADSA-N
1-palmitoyl-2-acetyl-gpc,pagpc,2s-2-acetyloxy-3-hexadecanoyloxypropyl 2-trimethylazaniumyl ethyl phosphate,3,5,9-trioxa-4-phosphapentacosan-1-aminium, 7-acetyloxy-4-hydroxy-n,n,n-trimethyl-10-oxo-, hydroxide, inner salt, 4-oxide, s,3,5,9-trioxa-4-phosphapentacosan-1-aminium,7-acetyloxy-4-hydroxy-n,n,n-trimethyl-10-oxo-,inner salt,4-oxide, s-
134690
[(2S)-2-acetyloxy-3-hexadecanoyloxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)C
84062-61-3
C26H52NO8P
537.675
MFCD00798242
XPAXRSJGGFVTFM-VWLOTQADSA-N
1-palmitoyl-2-acetyl-gpc,pagpc,2s-2-acetyloxy-3-hexadecanoyloxypropyl 2-trimethylazaniumyl ethyl phosphate,3,5,9-trioxa-4-phosphapentacosan-1-aminium, 7-acetyloxy-4-hydroxy-n,n,n-trimethyl-10-oxo-, hydroxide, inner salt, 4-oxide, s,3,5,9-trioxa-4-phosphapentacosan-1-aminium,7-acetyloxy-4-hydroxy-n,n,n-trimethyl-10-oxo-,inner salt,4-oxide, s-
134690
[(2S)-2-acetyloxy-3-hexadecanoyloxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)C
Quantity 1mg
Formula Weight 537.67
Physical Form Viscous Oil
Percent Purity 99%
Chemical Name or Material 1-O-Palmitoyl-2-O-acetyl-sn-glycero-3-phosphocholine

storageNote1 : -30° to -10°C