Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(o-Tolyl)piperazine, 97%

Click to view available options
Quantity:
5g
25g
100g
39512-51-1
C11H16N2
176.263
MFCD00040729
WICKLEOONJPMEQ-UHFFFAOYSA-N
1-2-methylphenyl piperazine,1-o-tolyl piperazine,4-2-methylphenyl piperazine,1-o-tolylpiperazine,piperazine, 1-2-methylphenyl,1-o-tolyl-piperazine,1-2-methylphenyl-piperazine,o-tolylpiperazine,2-methylphenylpiperazine,chembl9746
91965
1-(2-methylphenyl)piperazine
CC1=CC=CC=C1N2CCNCC2
39512-51-1
C11H16N2
176.263
MFCD00040729
WICKLEOONJPMEQ-UHFFFAOYSA-N
1-2-methylphenyl piperazine,1-o-tolyl piperazine,4-2-methylphenyl piperazine,1-o-tolylpiperazine,piperazine, 1-2-methylphenyl,1-o-tolyl-piperazine,1-2-methylphenyl-piperazine,o-tolylpiperazine,2-methylphenylpiperazine,chembl9746
91965
1-(2-methylphenyl)piperazine
CC1=CC=CC=C1N2CCNCC2
Quantity 5g
Melting Point 45°C to 48°C
UN Number UN3263
Formula Weight 176.26
Percent Purity 97%
Chemical Name or Material 1-(o-Tolyl)piperazine

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.