Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(o-Tolyl)piperazine dihydrochloride, 98%

Click to view available options
Quantity:
5g
25g
100g
70849-60-4
C11H17ClN2
212.721
MFCD00039030
LZNYIWJDVYCKDU-UHFFFAOYSA-N
1-o-tolyl piperazine hydrochloride,n-2-methylphenyl piperazine hydrochloride,1-2-methylphenyl piperazine hydrochloride,1-2-tolyl piperazine hydrochloride,piperazine, 1-2-methylphenyl-, monohydrochloride,piperazine, 1-2-methylphenyl-, hydrochloride,acmc-209ryj,1-o-tolyl piperazinehydrochloride,1-o-tolyl-piperazine hydrochloride,n-o-tolyl piperazine hydrochloride
12227895
1-(2-methylphenyl)piperazine;hydrochloride
CC1=CC=CC=C1N2CCNCC2.Cl
70849-60-4
C11H17ClN2
212.721
MFCD00039030
LZNYIWJDVYCKDU-UHFFFAOYSA-N
1-o-tolyl piperazine hydrochloride,n-2-methylphenyl piperazine hydrochloride,1-2-methylphenyl piperazine hydrochloride,1-2-tolyl piperazine hydrochloride,piperazine, 1-2-methylphenyl-, monohydrochloride,piperazine, 1-2-methylphenyl-, hydrochloride,acmc-209ryj,1-o-tolyl piperazinehydrochloride,1-o-tolyl-piperazine hydrochloride,n-o-tolyl piperazine hydrochloride
12227895
1-(2-methylphenyl)piperazine;hydrochloride
CC1=CC=CC=C1N2CCNCC2.Cl
Quantity 5g
Melting Point 238°C to 240°C
Beilstein 4019516
Formula Weight 249.19
Percent Purity 98%
Chemical Name or Material 1-(o-Tolyl)piperazine dihydrochloride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning