Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(o-Tolyl)piperazine Hydrochloride 97.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
95356-15-3
C11H17ClN2
212.721
MFCD00039030
LZNYIWJDVYCKDU-UHFFFAOYSA-N
1-(2-Methylphenyl)piperazine Hydrochloride
12227895
1-(2-methylphenyl)piperazine;hydrochloride
CC1=CC=CC=C1N2CCNCC2.Cl
95356-15-3
C11H17ClN2
212.721
MFCD00039030
LZNYIWJDVYCKDU-UHFFFAOYSA-N
1-(2-Methylphenyl)piperazine Hydrochloride
12227895
1-(2-methylphenyl)piperazine;hydrochloride
CC1=CC=CC=C1N2CCNCC2.Cl
Color White
Melting Point 240°C
Quantity 5g
Formula Weight 212.72
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (T)
Chemical Name or Material 1-(o-Tolyl)piperazine Hydrochloride