Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Octadecanesulfonic acid, sodium salt, 90%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
10g
Packaging:
Glass bottle
13893-34-0
C18H37NaO3S
356.54
MFCD00051526
KBAFDSIZQYCDPK-UHFFFAOYSA-M
sodium 1-octadecanesulfonate,sodium octadecane-1-sulfonate,sodium stearylsulfonate,sodium octadecanesulfonate,1-octadecanesulfonic acid sodium salt,sodium octadecane-1-sulphonate,1-octadecanesulfonic acid, sodium salt,sodium sterylsulfonate,sodium oxadecanesulfonate,sodium n-octadecylsulfonate
23668003
sodium;octadecane-1-sulfonate
CCCCCCCCCCCCCCCCCCS(=O)(=O)[O-].[Na+]
13893-34-0
C18H37NaO3S
356.54
MFCD00051526
KBAFDSIZQYCDPK-UHFFFAOYSA-M
sodium 1-octadecanesulfonate,sodium octadecane-1-sulfonate,sodium stearylsulfonate,sodium octadecanesulfonate,1-octadecanesulfonic acid sodium salt,sodium octadecane-1-sulphonate,1-octadecanesulfonic acid, sodium salt,sodium sterylsulfonate,sodium oxadecanesulfonate,sodium n-octadecylsulfonate
23668003
sodium;octadecane-1-sulfonate
CCCCCCCCCCCCCCCCCCS(=O)(=O)[O-].[Na+]
Melting Point 163°C to 165°C
Infrared Spectrum Authentic
Elemental Analysis 88% min. (C, H)
Loss on Drying 5% max. (1 G, 105°C, 1 Hrs, 760mm)
Color White
Packaging Glass bottle
Quantity 1g
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Linear Formula CH3(CH2)17SO3Na
Solubility Information Solubility in water: soluble
Formula Weight 356.54
Physical Form Powder
Percent Purity 90%
Chemical Name or Material 1-Octadecanesulfonic acid, sodium salt
Show More Show Less

EINECSNumber : 237-660-6