Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Octadecanol, 97%

Manufacturer:  Alfa Aesar A1202036

Quantity 500g

 View more versions of this product

Catalog No. AAA1202036


  Check Availability
Add to cart

Specifications

Description

Applications
1-Octadecanol has been used as surfactant for the functionalization of carbon nanotubes. It is employed as emulsion stabilizer for creams and lotions, quality modifier of lipsticks, additive for ointment base and cream conditioners for smooth touch to skin for cosmetic uses. It is auxiliary for emulsifiers.

Solubility
Insoluble in water. Soluble in alcohol, acetone, ether, benzene, chloroform.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a well ventilated place.
Description & Specifications

Chemical Identifiers

112-92-5
270.5
1362907
1-Octadecanol
CH3(CH2)17OH
MFCD00002823
14,8805

Specifications

112-92-5
CH3(CH2)17OH
MFCD00002823
14,8805
1-Octadecanol
97%
270.5
1362907
500g
97%

Safety and Handling

SDS & Safety Documents

RTECSNumber : RG2010000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit