Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Octanesulfonic acid, sodium salt, HPLC grade, Acros Organics

Click to view available options
Quantity:
5g
25g
100g
500g
5324-84-5
C8H17NaO3S
216.28
MFCD00007544
HRQDCDQDOPSGBR-UHFFFAOYSA-M
sodium 1-octanesulfonate,sodium octane-1-sulfonate,1-octanesulfonic acid sodium salt,sodium octanesulfonate,1-octanesulfonic acid, sodium salt,1-octanesulfonate, sodium,sodium octylsulfonate,unii-j46ez5jyb3,octyl sodium sulfonate,ipc-alks-8
23669624
sodium;octane-1-sulfonate
CCCCCCCCS(=O)(=O)[O-].[Na+]
5324-84-5
C8H17NaO3S
216.28
MFCD00007544
HRQDCDQDOPSGBR-UHFFFAOYSA-M
sodium 1-octanesulfonate,sodium octane-1-sulfonate,1-octanesulfonic acid sodium salt,sodium octanesulfonate,1-octanesulfonic acid, sodium salt,1-octanesulfonate, sodium,sodium octylsulfonate,unii-j46ez5jyb3,octyl sodium sulfonate,ipc-alks-8
23669624
sodium;octane-1-sulfonate
CCCCCCCCS(=O)(=O)[O-].[Na+]
For ion pair chromatography
Absorbance (0.25M aqueous solution),(0.25 M aqueous solution) at 0nm,0.04 max. at 260nm,0.05 max. at 240nm,0.05 max. at 250nm,0.075 max. at 230nm,0.1 max. at 220nm,0.2 max. at 210nm,0.3 max. at 200nm
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Melting Point >300.0°C
Water 2% max.
Color White
Linear Formula CH3(CH2)7SO3Na
Solubility Information Solubility in water: freely soluble. Other solubilities: 4g/L in methanol
Formula Weight 216.28
Physical Form Crystalline Powder
Percent Purity ≥98.0%
Chemical Name or Material 1-Octanesulfonic acid, sodium salt
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-195-4