Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Octanesulfonic acid, sodium salt monohydrate, 98%, ACROS Organics™
Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass Bottle
Specifications
Infrared Spectrum | Authentic |
Packaging | Glass Bottle |
Color | White |
Melting Point | >300.0°C |
Quantity | 100g |
Assay Percent Range | 98% |
Linear Formula | CH3(CH2)7SO3Na·H2O |
Beilstein | 04,III,25 |
Solubility Information | Solubility in water: soluble. Other solubilities: 4g/l methanol |
Formula Weight | 234.3 |
Show More |
Safety and Handling
GHS Signal Word: Warning