Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Octanesulfonic acid sodium salt, monohydrate, 99.7%, For HPLC analysis, MP Biomedicals™
Supplier: MP Biomedicals, Inc 0215177825
Description

Specifications
1-Octanesulfonic Acid, Sodium Salt, Monohydrate | |
Crystalline Powder | |
White |
Store at room temperature (15°C to 30°C) | |
25 g |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction