1-Octanol (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

Packaging Glass Bottle
Quantity 4L

 View more versions of this product

Catalog No. A402-4

  null
Add to Cart

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 111-87-5
    Molecular Formula C8H18O
    Linear Formula CH3(CH2)6CH2OH
    Formula Weight 130.23
    MDL Number MFCD00002988
    Synonym Caprylic Alcohol
    Chemical Name or Material 1-Octanol
    Grade Certified Grade

    Specifications

    Formula Weight 130.23
    Boiling Point 195°C
    Melting Point −16°C
    Quantity 4L
    Flash Point 81°C
    Physical Form Liquid
    Color Colorless
    Density 4.5 (Air = 1.0)
    Alpha Vector Octanol
    Molecular Formula C8H18O
    Free Acid 0.05% max.
    Identification Pass Test
    Packaging Glass Bottle
    Residue after Evaporation 0.02% max.
    MDL Number MFCD00002988
    CAS 111-87-5
    ChemAlert Storage Symbol Gray
    Synonym Caprylic Alcohol
    Additional Information Vapor Pressure: 0.03mbar at 20°C
    Boiling Range 2°C max.
    Linear Formula CH3(CH2)6CH2OH
    Chemical Name or Material 1-Octanol
    Grade Certified Grade
     

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit