Alfa Aesar™ 1-Octene, 97+%

Manufacturer: Alfa Aesar™ A11146AP

Quantity 500mL

 View more versions of this product

Catalog No. AAA11146AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 111-66-0
    Molecular Formula C8H16
    Formula Weight 112.22
    MDL Number MFCD00009548
    UN Number UN3295
    Beilstein 1734497
    Merck Index 14,1764
    Synonym Caprylene; Octylene
    Chemical Name or Material 1-Octene

    Specifications

    Formula Weight 112.22
    Health Hazard 3 GHS P Statement
    P210-P280g-P273-P243-P301+P330+P331-P315
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves.
    Avoid release to the environment.
    Take precautionary measures against static discharge.
    IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
    Get immediate medical advice/attention.
    MDL Number MFCD00009548
    CAS 111-66-0
    UN Number UN3295
    Health Hazard 1 Danger
    Boiling Point 122° to 123°C
    TSCA TSCA
    Molecular Formula C8H16
    Merck Index 14,1764
    Quantity 500mL
    Health Hazard 2 GHS H Statement
    H225-H304-H315
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Causes skin irritation.
    Odor Petroleum-like
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
    RTECS Number RH2207000
    Beilstein 1734497
    Synonym Caprylene; Octylene
    Melting Point −102°C
    Flash Point 21°C (70°F)
    Chemical Name or Material 1-Octene
    Assay ≥97%

    Description

    Applications
    1-Octene acts as a comonomer used in the preparation of polyethylene, especially high-density polyethylene (HDPE) and linear low-density polyethylene(LLDPE) resins. It is also used in organic synthesis, surfactants and plasticizers. Further, it is used in process regulators and viscosity adjustors. In addition to this, it is used in the preparation of linear aldehyde through the oxo synthesis (hydroformylation) to get the nonanal.

    Solubility
    Miscible with water, ether, alcohol and acetone.

    Notes
    Incompatible with bromine, chlorine, acids and oxidizing agents.

    GHS H Statement
    H225-H304-H315
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Causes skin irritation.

    GHS P Statement
    P210-P280g-P273-P243-P301+P330+P331-P315
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves.
    Avoid release to the environment.
    Take precautionary measures against static discharge.
    IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
    Get immediate medical advice/attention.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit