Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Octylamine, 99%

Click to view available options
Quantity:
100g
500g
111-86-4
C8H19N
129.247
MFCD00008247
IOQPZZOEVPZRBK-UHFFFAOYSA-N
octylamine,n-octylamine,1-aminooctane,1-octanamine,1-octylamine,caprylamine,caprylylamine,armeen 8,n-octylamine, mono,armeen 8d
8143
CHEBI:7728
octan-1-amine
CCCCCCCCN
111-86-4
C8H19N
129.247
MFCD00008247
IOQPZZOEVPZRBK-UHFFFAOYSA-N
octylamine,n-octylamine,1-aminooctane,1-octanamine,1-octylamine,caprylamine,caprylylamine,armeen 8,n-octylamine, mono,armeen 8d
8143
CHEBI:7728
octan-1-amine
CCCCCCCCN
Applications
1-Octylamine is used as a precursor to prepare chemicals. It finds application in functional fluids and as a laboratory reagent. Further, it is used as a reactant in the preparation of 2H-indazoles and 1H-indazolones, which are used as myeloperoxidase(MPO) inhibitors.

Solubility
Miscible with ethanol, benzene and ethyl acetate. Slightly miscible with water.

Notes
Incompatible with acids, strong oxidizing agents, acid anhydrides and acid chlorides.
Melting Point -1°C
Boiling Point 178°C to 179°C
Quantity 100g
Flash Point 62°C (144°F)
UN Number UN2735
Beilstein 1679227
Formula Weight 129.25
Percent Purity 99%
Chemical Name or Material 1-Octylamine

GHS H Statement
H314-H318-H302-H312-H332-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

RTECSNumber : RG8050000

TSCA : TSCA