Alfa Aesar™ 1-Octylmagnesium chloride, 1M in MeTHF

Manufacturer: Alfa Aesar™ H51174AP

Quantity 500mL

 View more versions of this product

Catalog No. AAH51174AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 38841-98-4
    Molecular Formula C8H17ClMg
    Linear Formula CH3(CH2)6CH2MgCl
    Formula Weight 172.98
    MDL Number MFCD00000476
    UN Number UN2924
    Chemical Name or Material 1-Octylmagnesium chloride

    Specifications

    Formula Weight 172.98
    TSCA TSCA
    Molecular Formula C8H17ClMg
    Linear Formula CH3(CH2)6CH2MgCl
    MDL Number MFCD00000476
    CAS 38841-98-4
    UN Number UN2924
    Health Hazard 1 Danger
    Concentration 1M in MeTHF
    Quantity 500mL
    Health Hazard 2 GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Chemical Name or Material 1-Octylmagnesium chloride

    GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit