Learn More
Alfa Aesar™ 1-Octylmagnesium chloride, 1M in MeTHF
Manufacturer:  Alfa Aesar™ H51174AE
Quantity 100mL
 View more versions of this product
Catalog No. AAH51174AE
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Chemical Identifiers
38841-98-4
CH3(CH2)6CH2MgCl
MFCD00000476
1-Octylmagnesium chloride
C8H17ClMg
172.98
UN2924
Specifications
38841-98-4
CH3(CH2)6CH2MgCl
MFCD00000476
1M in MeTHF
1-Octylmagnesium chloride
C8H17ClMg
172.98
UN2924
100mL
Safety and Handling
SDS & Safety Documents
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA : TSCA