Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-(p-Toluenesulfonyl)indole, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
1-(p-Toluenesulfonyl)indole, is synthesized and used as a stable equivalent of 4-mercaptoindoles, also used in medicinal field for making drug and tablets.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-(p-Toluenesulfonyl)indole, is synthesized and used as a stable equivalent of 4-mercaptoindoles, also used in medicinal field for making drug and tablets.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
1-(p-Toluenesulfonyl)indole, is synthesized and used as a stable equivalent of 4-mercaptoindoles, also used in medicinal field for making drug and tablets.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 84°C to 86°C |
Quantity | 25 g |
Solubility Information | Insoluble in water. |
IUPAC Name | 1-(4-methylphenyl)sulfonylindole |
Formula Weight | 271.33 |
Percent Purity | 95% |
Chemical Name or Material | 1-(p-Toluenesulfonyl)indole |
Safety and Handling
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction