Alfa Aesar™ 1-Pentadecene, 97%

Manufacturer: Alfa Aesar™ H5351614

Quantity 25g

 View more versions of this product

Catalog No. AAH5351614

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 13360-61-7
    Molecular Formula C15H30
    Linear Formula CH3(CH2)12CH=CH2
    Formula Weight 210.4
    MDL Number MFCD00008987
    Beilstein 1751011
    Chemical Name or Material 1-Pentadecene

    Specifications

    Formula Weight 210.4
    TSCA TSCA
    Linear Formula CH3(CH2)12CH=CH2
    CAS 13360-61-7
    Melting Point −4°C
    Quantity 25g
    Molecular Formula C15H30
    MDL Number MFCD00008987
    Beilstein 1751011
    Boiling Point 268° to 269°C
    Flash Point 110°C (230°F)
    Chemical Name or Material 1-Pentadecene
    Assay 97%

    Description

    Applications
    1-Pentadecene serves as a growth medium for the strain CV2803T. It is a common semiochemical of flour beetles.

    Solubility
    Not miscible or difficult to mix in water.

    Notes
    Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit