Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pentanesulfonic acid, sodium salt, 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
22767-49-3
C5H11NaO3S
174.19
MFCD00007541
ROBLTDOHDSGGDT-UHFFFAOYSA-M
sodium 1-pentanesulfonate,sodium pentane-1-sulfonate,sodium pentanesulfonate,1-pentanesulfonic acid sodium salt,1-pentanesulfonic acid, sodium salt,1-pentylsodiumsulfonate,ipc-alks-5,1-pentanesulfonate, sodium,pentanesulfonic acid na-salt,pentanesulfonic acidsodiumsalt
23664617
sodium;pentane-1-sulfonate
CCCCCS(=O)(=O)[O-].[Na+]
22767-49-3
C5H11NaO3S
174.19
MFCD00007541
ROBLTDOHDSGGDT-UHFFFAOYSA-M
sodium 1-pentanesulfonate,sodium pentane-1-sulfonate,sodium pentanesulfonate,1-pentanesulfonic acid sodium salt,1-pentanesulfonic acid, sodium salt,1-pentylsodiumsulfonate,ipc-alks-5,1-pentanesulfonate, sodium,pentanesulfonic acid na-salt,pentanesulfonic acidsodiumsalt
23664617
sodium;pentane-1-sulfonate
CCCCCS(=O)(=O)[O-].[Na+]
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point >300.0°C
Quantity 5g
Color White to Yellow
Loss on Drying 2 % max. (1 g, 120°C)
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula CH3(CH2)4SO3Na
Beilstein 04, III, 23
Solubility Information Solubility in water: soluble.
Formula Weight 174.2
Physical Form Crystals
Percent Purity ≥98%
Chemical Name or Material 1-Pentanesulfonic acid, sodium salt
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.