Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pentanesulfonic Acid Sodium Salt, HPLC Grade, J.T.Baker™

Click to view available options
Quantity:
25g
100g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

22767-49-3
C5H11NaO3S
174.19
ROBLTDOHDSGGDT-UHFFFAOYSA-M
23664617

High purity solvents for use in analytical and preparative separations

    For Ion-Pair Chromatography of Basic Compounds; For Laboratory,Research,or Manufacturing Use
Quantity 100g
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 0.00kg
SMILES CCCCCS(=O)(=O)[O-].[Na+]
IUPAC Name sodium;pentane-1-sulfonate
Formula Weight 174.21
Chemical Name or Material 1-Pentanesulfonic Acid Sodium Salt
Grade HPLC Grade