Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pentanesulfonic Acid Sodium Salt, Monohydrate, HPLC Grade, 98%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
207605-40-1
C5H11O3S
151.20
RJQRCOMHVBLQIH-UHFFFAOYSA-M
pentane-1-sulfonate
CCCCCS([O-])(=O)=O
207605-40-1
C5H11O3S
151.20
RJQRCOMHVBLQIH-UHFFFAOYSA-M
pentane-1-sulfonate
CCCCCS([O-])(=O)=O

P1043, 207605-40-1, C5H11O3SNa•H2O

Spectrum™ 1-Pentanesulfonic Acid Sodium Salt, Monohydrate, HPLC Grade
Packaging Amber Glass Bottle
Quantity 1 kg
CAS Max % 1
Percent Purity 98%