Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pentanol, Reagent, ACS, 98%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
71-41-0
C5H12O
88.15
AMQJEAYHLZJPGS-UHFFFAOYSA-N
pentan-1-ol
CCCCCO
71-41-0
C5H12O
88.15
AMQJEAYHLZJPGS-UHFFFAOYSA-N
pentan-1-ol
CCCCCO

A1285, 71-41-0, C5H12O

Spectrum™ 1-Pentanol, Reagent, ACS
Packaging Amber Glass Bottle
Residue after Evaporation 3e-005
Water 0.005
Quantity 4 L
CAS Max % 1
UN Number UN1105
Percent Purity 98%