Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Penten-3-ol, 97%, Acros Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
616-25-1
C5H10O
86.13
MFCD00004573
VHVMXWZXFBOANQ-UHFFFAOYSA-N
1-penten-3-ol,ethyl vinyl carbinol,vinyl ethyl carbinol,1-pentene-3-ol,1-ethylallyl alcohol,alpha-ethylallyl alcohol,1-penten-3-ol e,1-penten-3-ol, r,fema no. 3584,ethylvinylcarbinol
12020
pent-1-en-3-ol
CCC(C=C)O
616-25-1
C5H10O
86.13
MFCD00004573
VHVMXWZXFBOANQ-UHFFFAOYSA-N
1-penten-3-ol,ethyl vinyl carbinol,vinyl ethyl carbinol,1-pentene-3-ol,1-ethylallyl alcohol,alpha-ethylallyl alcohol,1-penten-3-ol e,1-penten-3-ol, r,fema no. 3584,ethylvinylcarbinol
12020
pent-1-en-3-ol
CCC(C=C)O
Refractive Index 1.4220 to 1.4260
Specific Gravity 0.839
Packaging Glass bottle
Quantity 25g
Color Yellow to Beige
Density 0.8390g/mL
Flash Point 25°C
Infrared Spectrum Authentic
Boiling Point 114.0°C to 115.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C2H5CH(OH)CH=CH2
Beilstein 01,443
Solubility Information Solubility in water: slightly soluble in water
Formula Weight 86.13
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 1-Penten-3-ol, 99%
Show More Show Less

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 210-472-1

TSCA : TSCA