Alfa Aesar™ 1-Pentylmagnesium chloride, 1M in MeTHF

Manufacturer: Alfa Aesar™ H51175AE

Quantity 100mL

 View more versions of this product

Catalog No. AAH51175AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6393-56-2
    Molecular Formula C5H11ClMg
    Linear Formula CH3(CH2)4MgCl
    Formula Weight 130.9
    MDL Number MFCD00672089
    UN Number UN2924
    Chemical Name or Material 1-Pentylmagnesium chloride

    Specifications

    Formula Weight 130.9
    Concentration 1M in MeTHF
    Molecular Formula C5H11ClMg
    Quantity 100mL
    Linear Formula CH3(CH2)4MgCl
    MDL Number MFCD00672089
    CAS 6393-56-2
    UN Number UN2924
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Chemical Name or Material 1-Pentylmagnesium chloride

    GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit