Learn More
Alfa Aesar™ 1-Pentylmagnesium chloride, 1M in MeTHF
Manufacturer:  Alfa Aesar™ H51175AP
Quantity 500mL
 View more versions of this product
Catalog No. AAH51175AP
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
6393-56-2
CH3(CH2)4MgCl
MFCD00672089
1-Pentylmagnesium chloride
C5H11ClMg
130.9
UN2924
Specifications
6393-56-2
C5H11ClMg
CH3(CH2)4MgCl
MFCD00672089
1-Pentylmagnesium chloride
1M in MeTHF
500mL
130.9
UN2924
Safety and Handling
SDS & Safety Documents
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.