Alfa Aesar™ 1-Pentyn-3-ol, 98%

Manufacturer: Alfa Aesar™ B2485303

Quantity 1g

 View more versions of this product

Catalog No. AAB2485303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4187-86-4
    Molecular Formula C5H8O
    Formula Weight 84.12
    MDL Number MFCD00004572
    UN Number UN1986
    Beilstein 1098409
    Synonym Ethyl ethynyl carbinol
    Chemical Name or Material 1-Pentyn-3-ol

    Specifications

    Formula Weight 84.12
    Boiling Point 123° to 125°C
    Quantity 1g
    Flash Point 29°C (84°F)
    UN Number UN1986
    Beilstein 1098409
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P260-P280f-P307+P311
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective clothing.
    IF exposed: Call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, FLAMMABLE, TOXIC, N.O.S.
    Odor Recognizable
    RTECS Number SC4758500
    Molecular Formula C5H8O
    MDL Number MFCD00004572
    CAS 4187-86-4
    Synonym Ethyl ethynyl carbinol
    Health Hazard 2 GHS H Statement
    H301-H226
    Toxic if swallowed.
    Flammable liquid and vapour.
    Chemical Name or Material 1-Pentyn-3-ol
    Assay 98%

    Description

    Applications
    1-Pentyn-3-ol is used as a pharmaceutical intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents, heavy metal powders.

    GHS H Statement
    H301-H226
    Toxic if swallowed.
    Flammable liquid and vapour.

    GHS P Statement
    P260-P280f-P307+P311
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective clothing.
    IF exposed: Call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit