Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-1,2-ethanediol, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
Plastic bottle
93-56-1
C8H10O2
138.17
MFCD00003546
PWMWNFMRSKOCEY-UHFFFAOYSA-N
1-phenyl-1,2-ethanediol,styrene glycol,phenylethylene glycol,phenylethanediol,phenyl glycol,1,2-ethanediol, 1-phenyl,phenyl-1,2-ethanediol,styrolyl alcohol,fenylglycol,1-phenylethylene glycol
7149
1-phenylethane-1,2-diol
C1=CC=C(C=C1)C(CO)O
93-56-1
C8H10O2
138.17
MFCD00003546
PWMWNFMRSKOCEY-UHFFFAOYSA-N
1-phenyl-1,2-ethanediol,styrene glycol,phenylethylene glycol,phenylethanediol,phenyl glycol,1,2-ethanediol, 1-phenyl,phenyl-1,2-ethanediol,styrolyl alcohol,fenylglycol,1-phenylethylene glycol
7149
1-phenylethane-1,2-diol
C1=CC=C(C=C1)C(CO)O
Boiling Point 272°C to 274°C
Packaging Plastic bottle
Infrared Spectrum Authentic
Melting Point 61°C to 67°C
Quantity 25g
Color White to Beige
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula HOCH2CH(C6H5)OH
Beilstein 06, 907
Merck Index 15, 8991
Formula Weight 138.17
Physical Form Crystalline Powder or Flakes
Percent Purity ≥96.0%
Chemical Name or Material 1-Phenyl-1, 2-ethanediol
Show More Show Less