Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-1,2-propanedione-2-oxime, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
Packaging:
Glass bottle
119-51-7
C9H9NO2
163.18
MFCD00002115
YPINLRNGSGGJJT-JXMROGBWSA-N
1-phenyl-1,2-propanedione-2-oxime,2-hydroxyimino propiophenone,isonitrosopropiophenone,2-hydroxyiminopropiophenone,2-hydroxyimino-1-phenylpropan-1-one,propiophenone, isonitroso,2-isonitrosopropiophenone,alpha-oximinopropiophenone,1-phenyl-1,2-propanedione 2-oxime,.alpha.-oximinopropiophenone
9566063
(2E)-2-hydroxyimino-1-phenylpropan-1-one
CC(=NO)C(=O)C1=CC=CC=C1
119-51-7
C9H9NO2
163.18
MFCD00002115
YPINLRNGSGGJJT-JXMROGBWSA-N
1-phenyl-1,2-propanedione-2-oxime,2-hydroxyimino propiophenone,isonitrosopropiophenone,2-hydroxyiminopropiophenone,2-hydroxyimino-1-phenylpropan-1-one,propiophenone, isonitroso,2-isonitrosopropiophenone,alpha-oximinopropiophenone,1-phenyl-1,2-propanedione 2-oxime,.alpha.-oximinopropiophenone
9566063
(2E)-2-hydroxyimino-1-phenylpropan-1-one
CC(=NO)C(=O)C1=CC=CC=C1
Quantity 10g
Melting Point 111.0°C to 115.0°C
Color White to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H5COC(=NOH)CH3
Beilstein 07, III, 3464
Solubility Information Solubility in water: 1.3g/L (20°C)
Formula Weight 163.18
Physical Form Crystals
Percent Purity 99%
Chemical Name or Material 1-Phenyl-1, 2-propanedione-2-oxime, 99%
Show More Show Less

EINECSNumber : 204-329-2

RTECSNumber : UH2975000

TSCA : TSCA