Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Phenyl-1,2-propanedione-2-oxime, 99%, ACROS Organics™
Click to view available options
Quantity:
10g
25g
Packaging:
Glass bottle
Specifications
Packaging | Glass bottle |
Color | White to Yellow |
Melting Point | 111.0°C to 115.0°C |
Quantity | 25g |
Infrared Spectrum | Authentic |
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Assay Percent Range | 99% |
Linear Formula | C6H5COC(=NOH)CH3 |
Beilstein | 07, III, 3464 |
Show More |
Safety and Handling
EINECSNumber : 204-329-2
RTECSNumber : UH2975000
TSCA : TSCA