Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-1-butyne, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
622-76-4
C10H10
130.19
MFCD00039945
FFFMSANAQQVUJA-UHFFFAOYSA-N
1-phenyl-1-butyne,1-butynylbenzene,benzene, 1-butynyl,1-butyne, 1-phenyl,but-1-yn-1-ylbenzene,1-but-1-ynyl benzene,1-ethyl-2-phenylacetylene,phenylbutyne,ethylphenylacetylene,phenylethylacetylene
69328
but-1-ynylbenzene
CCC#CC1=CC=CC=C1
622-76-4
C10H10
130.19
MFCD00039945
FFFMSANAQQVUJA-UHFFFAOYSA-N
1-phenyl-1-butyne,1-butynylbenzene,benzene, 1-butynyl,1-butyne, 1-phenyl,but-1-yn-1-ylbenzene,1-but-1-ynyl benzene,1-ethyl-2-phenylacetylene,phenylbutyne,ethylphenylacetylene,phenylethylacetylene
69328
but-1-ynylbenzene
CCC#CC1=CC=CC=C1
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Colorless
Density 0.916g/mL
Refractive Index 1.5500 to 1.5520 (20°C, 589nm)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H5C2CCH2CH3
Formula Weight 130.19
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 1-Phenyl-1-butyne
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.