Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-1-pentanol 98.0+%, TCI America™

Click to view available options
Quantity:
5mL
25mL
583-03-9
C11H16O
164.248
MFCD00021935
OVGORFFCBUIFIA-UHFFFAOYSA-N
fenipentol, 1-phenyl-1-pentanol, 1-phenylpentanol, pancoral, phenylbutylcarbinol, suiclisin, alpha-butylbenzyl alcohol, 1-pentanol, 1-phenyl, 1-phenyl-1-hydroxypentane, ph bc
3338
1-phenylpentan-1-ol
CCCCC(C1=CC=CC=C1)O
583-03-9
C11H16O
164.248
MFCD00021935
OVGORFFCBUIFIA-UHFFFAOYSA-N
fenipentol, 1-phenyl-1-pentanol, 1-phenylpentanol, pancoral, phenylbutylcarbinol, suiclisin, alpha-butylbenzyl alcohol, 1-pentanol, 1-phenyl, 1-phenyl-1-hydroxypentane, ph bc
3338
1-phenylpentan-1-ol
CCCCC(C1=CC=CC=C1)O
Color Colorless
Boiling Point 137°C
Quantity 25mL
Formula Weight 164.25
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 1-Phenyl-1-pentanol

RTECSNumber : DN8780000

TSCA : Yes