Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (+/-)-1-Phenyl-1-propanol, 98+%

Manufacturer:  Alfa Aesar™ A1083036

Quantity 500g

 View more versions of this product

Catalog No. AAA1083036


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

93-54-9
136.19
1906759
(+/-)-alpha-Ethylbenzyl alcohol; Phenyl ethyl carbinol
C9H12O
MFCD00004564
14,3768
(±)-1-Phenyl-1-propanol

Specifications

500g
C9H12O
MFCD00004564
14,3768
217° to 219°C
(±)-1-Phenyl-1-propanol
93-54-9
136.19
1906759
(+/-)-alpha-Ethylbenzyl alcohol; Phenyl ethyl carbinol
95°C (203°F)
≥98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DO5470000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit