Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-1-propanol 99%, ACROS Organics™

Click to view available options
Quantity:
50g
Packaging:
Glass bottle
93-54-9
C9H12O
136.194
MFCD00004564
DYUQAZSOFZSPHD-UHFFFAOYSA-N
1-phenyl-1-propanol,phenycholon,fepar,ejibil,felicur,livonal,1-phenylpropanol,gallenperlen,phenylchol,bilergon
7147
1-phenylpropan-1-ol
CCC(C1=CC=CC=C1)O
93-54-9
C9H12O
136.194
MFCD00004564
DYUQAZSOFZSPHD-UHFFFAOYSA-N
1-phenyl-1-propanol,phenycholon,fepar,ejibil,felicur,livonal,1-phenylpropanol,gallenperlen,phenylchol,bilergon
7147
1-phenylpropan-1-ol
CCC(C1=CC=CC=C1)O
Packaging Glass bottle
Refractive Index 1.5190 to 1.5210 (20°C, 589nm)
Quantity 50g
Density 0.9900g/mL
Color Colorless
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C2H5CH(C6H5)OH
Formula Weight 136.19
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 1-Phenyl-1-propanol, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 202-256-0

RTECSNumber : DO5470000

TSCA : TSCA