Alfa Aesar™ 1-Phenyl-2-hexanone, 95%

Manufacturer: Alfa Aesar™ A1094406

Quantity 5g

 View more versions of this product

Catalog No. AAA1094406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 25870-62-6
    Molecular Formula C12H16O
    Formula Weight 176.26
    MDL Number MFCD00027240
    Beilstein 2044423
    Synonym Benzyl n-butyl ketone
    Chemical Name or Material 1-Phenyl-2-hexanone

    Specifications

    Formula Weight 176.26
    Quantity 5g
    Molecular Formula C12H16O
    MDL Number MFCD00027240
    CAS 25870-62-6
    Beilstein 2044423
    Synonym Benzyl n-butyl ketone
    Boiling Point 153°C (20mmHg)
    Flash Point >110°C (230°F)
    Chemical Name or Material 1-Phenyl-2-hexanone
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit