Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-2-propanol, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
14898-87-4
C9H12O
136.19
MFCD00004540
WYTRYIUQUDTGSX-UHFFFAOYNA-N
1-phenyl-2-propanol, benzyl methyl carbinol, 2-hydroxy-1-phenylpropane, benzylmethylcarbinol, 2-propanol, 1-phenyl, benzeneethanol, .alpha.-methyl, alpha-methyl-phenethyl alcohol, benzeneethanol, alpha-methyl, alpha-methylbenzeneethanol, .alpha.-methylphenethyl alcohol
94185
1-phenylpropan-2-ol
CC(O)CC1=CC=CC=C1
14898-87-4
C9H12O
136.19
MFCD00004540
WYTRYIUQUDTGSX-UHFFFAOYNA-N
1-phenyl-2-propanol, benzyl methyl carbinol, 2-hydroxy-1-phenylpropane, benzylmethylcarbinol, 2-propanol, 1-phenyl, benzeneethanol, .alpha.-methyl, alpha-methyl-phenethyl alcohol, benzeneethanol, alpha-methyl, alpha-methylbenzeneethanol, .alpha.-methylphenethyl alcohol
94185
1-phenylpropan-2-ol
CC(O)CC1=CC=CC=C1
Specific Gravity 0.99
Flash Point 85°C
Packaging Glass bottle
Refractive Index 1.5200 to 1.522
Color Colorless
Infrared Spectrum Authentic
Quantity 5g
Density 0.9900g/mL
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula C6H5CH2CH(OH)CH3
Beilstein 06,503
Formula Weight 136.19
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 1-Phenyl-2-propanol
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning