Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-2-propanol, 99%, Thermo Scientific Chemicals

Catalog Number AC183060250
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
14898-87-4
C9H12O
136.19
MFCD00004540
WYTRYIUQUDTGSX-UHFFFAOYNA-N
1-phenyl-2-propanol, benzyl methyl carbinol, 2-hydroxy-1-phenylpropane, benzylmethylcarbinol, 2-propanol, 1-phenyl, benzeneethanol, .alpha.-methyl, alpha-methyl-phenethyl alcohol, benzeneethanol, alpha-methyl, alpha-methylbenzeneethanol, .alpha.-methylphenethyl alcohol
94185
1-phenylpropan-2-ol
CC(O)CC1=CC=CC=C1
14898-87-4
C9H12O
136.19
MFCD00004540
WYTRYIUQUDTGSX-UHFFFAOYNA-N
1-phenyl-2-propanol, benzyl methyl carbinol, 2-hydroxy-1-phenylpropane, benzylmethylcarbinol, 2-propanol, 1-phenyl, benzeneethanol, .alpha.-methyl, alpha-methyl-phenethyl alcohol, benzeneethanol, alpha-methyl, alpha-methylbenzeneethanol, .alpha.-methylphenethyl alcohol
94185
1-phenylpropan-2-ol
CC(O)CC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.9900g/mL
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5200 to 1.522
Linear Formula C6H5CH2CH(OH)CH3
Quantity 25 g
Beilstein 06,503
Specific Gravity 0.99
Formula Weight 136.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Phenyl-2-propanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit