Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-2-thiourea, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
103-85-5
C7H8N2S
152.21
MFCD00004933
FULZLIGZKMKICU-UHFFFAOYSA-N
n-phenylthiourea,1-phenylthiourea,1-phenyl-2-thiourea,phenylthiocarbamide,thiourea, phenyl,phenyl-2-thiourea,fenylthiomocovina,urea, 1-phenyl-2-thio,rcra waste number p093,phenyl-thiourea
676454
CHEBI:46261
phenylthiourea
C1=CC=C(C=C1)NC(=S)N
103-85-5
C7H8N2S
152.21
MFCD00004933
FULZLIGZKMKICU-UHFFFAOYSA-N
n-phenylthiourea,1-phenylthiourea,1-phenyl-2-thiourea,phenylthiocarbamide,thiourea, phenyl,phenyl-2-thiourea,fenylthiomocovina,urea, 1-phenyl-2-thio,rcra waste number p093,phenyl-thiourea
676454
CHEBI:46261
phenylthiourea
C1=CC=C(C=C1)NC(=S)N
Color White to Beige
Melting Point 148°C to 150°C
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Linear Formula C6H5NHCSNH2
Beilstein 12, 388
Merck Index 15, 7424
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: soluble in alcohol,aq.ether,naoh,ethanol
Formula Weight 152.21
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 1-Phenyl-2-thiourea
Show More Show Less

GHS H Statement
May cause an allergic skin reaction.
Fatal if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTE

GHS Signal Word: Danger

EINECSNumber : 203-151-2