Alfa Aesar™ 1-Phenyl-2-trimethylsilylacetylene, 99%

Manufacturer: Alfa Aesar™ H5586606

Quantity 5g

 View more versions of this product

Catalog No. AAH5586606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2170-06-1
    Linear Formula C6H5C≡CSi(CH3)3
    Formula Weight 174.32
    MDL Number MFCD00054860
    Beilstein 3088678
    Synonym Phenylethynyltrimethylsilane; Trimethyl(phenylethynyl)silane
    Chemical Name or Material 1-Phenyl-2-trimethylsilylacetylene

    Specifications

    Formula Weight 174.32
    Boiling Point 87° to 88°C (9mmHg)
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    Synonym Phenylethynyltrimethylsilane; Trimethyl(phenylethynyl)silane
    Health Hazard 1 Warning
    Density 0.886g/mL
    Color Colorless
    Quantity 5g
    MDL Number MFCD00054860
    CAS 2170-06-1
    Beilstein 3088678
    Refractive Index 1.5280
    Linear Formula C6H5C≡CSi(CH3)3
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Chemical Name or Material 1-Phenyl-2-trimethylsilylacetylene
    Assay 99%

    Description

    Applications
    1-Phenyl-2-trimethylsilylacetylene was used in the preparation of donor-stabilized Pt-η-alkyne complexes.

    Solubility
    Sparingly Soluble (6.1E-3 g/L) (25°C)

    Notes
    Store at room temperature. Avoid strong oxidizing agents.

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit