Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-3-ethyl-1-heptyn-3-ol, 98%

Click to view available options
Quantity:
5g
19781-33-0
C15H20O
216.324
MFCD00041580
MIVGYFVSMBKLHD-UHFFFAOYSA-N
1-phenyl-3-ethyl-1-heptyn-3-ol,acmc-20aosc,1-phenyl-3-ethyl-1-heptyne-3-ol,1-heptyn-3-ol,3-ethyl-1-phenyl
20382016
3-ethyl-1-phenylhept-1-yn-3-ol
CCCCC(CC)(C#CC1=CC=CC=C1)O
19781-33-0
C15H20O
216.324
MFCD00041580
MIVGYFVSMBKLHD-UHFFFAOYSA-N
1-phenyl-3-ethyl-1-heptyn-3-ol,acmc-20aosc,1-phenyl-3-ethyl-1-heptyne-3-ol,1-heptyn-3-ol,3-ethyl-1-phenyl
20382016
3-ethyl-1-phenylhept-1-yn-3-ol
CCCCC(CC)(C#CC1=CC=CC=C1)O
Quantity 5g
Formula Weight 216.33
Percent Purity 98%
Chemical Name or Material 1-Phenyl-3-ethyl-1-heptyn-3-ol