Alfa Aesar™ 1-Phenyl-3-pyrazolidinone, 97%

Manufacturer: Alfa Aesar™ A1208918

Quantity 50g

 View more versions of this product

Catalog No. AAA1208918

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 92-43-3
    Molecular Formula C9H10N2O
    Formula Weight 162.19
    MDL Number MFCD00003094
    UN Number UN2811
    Beilstein 131856
    Merck Index 14,7309
    Synonym Phenidone
    Chemical Name or Material 1-Phenyl-3-pyrazolidinone

    Specifications

    Formula Weight 162.19
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    TSCA TSCA
    CAS 92-43-3
    Synonym Phenidone
    Melting Point 120° to 123°C
    Molecular Formula C9H10N2O
    MDL Number MFCD00003094
    Merck Index 14,7309
    Health Hazard 1 Warning
    UN Number UN2811
    Beilstein 131856
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    RTECS Number UQ8750000
    Quantity 50g
    Chemical Name or Material 1-Phenyl-3-pyrazolidinone
    Assay 97%

    Description

    Applications
    Phenidone inhibits both the LO (lipoxygenase) and Cox (cyclooxygenase) pathways, the synthesis of Fos-related antigen protein, and is described as an anti-inflammatory and anti-oxidant compound. Phenidone has been shown to block neurotoxicity induced by kainate, and weakens oxidative stress induced by lipopolysaccharide.

    Solubility
    Soluble in water (1:10 hot), methanol (0.1 mg/mL), DMSO (75 mg/mL), and ethanol.

    Notes
    Store in cool place. Keep away from oxidizing agents.

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit