Alfa Aesar™ 1-Phenyl-3-trimethylsilyl-2-propyn-1-ol, 97%

Manufacturer: Alfa Aesar™ H5342606

Quantity 5g

 View more versions of this product

Catalog No. AAH5342606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 89530-34-7
    Molecular Formula C12H16OSi
    Formula Weight 204.34
    MDL Number MFCD05864336
    Synonym alpha-[(Trimethylsilyl)ethynyl]benzenemethanol
    Chemical Name or Material 1-Phenyl-3-trimethylsilyl-2-propyn-1-ol

    Specifications

    Formula Weight 204.34
    Molecular Formula C12H16OSi
    Quantity 5g
    Flash Point 110°C (230°F)
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    MDL Number MFCD05864336
    CAS 89530-34-7
    Synonym alpha-[(Trimethylsilyl)ethynyl]benzenemethanol
    Health Hazard 2 GHS H Statement
    H319
    Causes serious eye irritation.
    Chemical Name or Material 1-Phenyl-3-trimethylsilyl-2-propyn-1-ol
    Assay 97%

    GHS H Statement
    H319
    Causes serious eye irritation.

    GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit