Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-4,5-dichloro-6-pyridazone, 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
1698-53-9
C10H6Cl2N2O
241.071
MFCD00006470
VKWCOHVAHQOJGU-UHFFFAOYSA-N
4,5-dichloro-2-phenylpyridazin-3 2h-one,1-phenyl-4,5-dichloro-6-pyridazone,3 2h-pyridazinone, 4,5-dichloro-2-phenyl,4,5-dichloro-2-phenyl-3 2h-pyridazinone,1-fenyl-4,5-dichlor-6-pyridazinon,2-phenyl-4,5-dichloro-3-pyridazinone,4,5-dichloro-2,3-dihydro-2-phenylpyridazin-3-one,4,5-dichloro-2-phenyl-2,3-dihydropyridazin-3-one,1-fenyl-4,5-dichlor-6-pyridazinon czech,4,5-dichloro-2-phenyl-pyridazin-3-one
72813
4,5-dichloro-2-phenylpyridazin-3-one
C1=CC=C(C=C1)N2C(=O)C(=C(C=N2)Cl)Cl
1698-53-9
C10H6Cl2N2O
241.071
MFCD00006470
VKWCOHVAHQOJGU-UHFFFAOYSA-N
4,5-dichloro-2-phenylpyridazin-3 2h-one,1-phenyl-4,5-dichloro-6-pyridazone,3 2h-pyridazinone, 4,5-dichloro-2-phenyl,4,5-dichloro-2-phenyl-3 2h-pyridazinone,1-fenyl-4,5-dichlor-6-pyridazinon,2-phenyl-4,5-dichloro-3-pyridazinone,4,5-dichloro-2,3-dihydro-2-phenylpyridazin-3-one,4,5-dichloro-2-phenyl-2,3-dihydropyridazin-3-one,1-fenyl-4,5-dichlor-6-pyridazinon czech,4,5-dichloro-2-phenyl-pyridazin-3-one
72813
4,5-dichloro-2-phenylpyridazin-3-one
C1=CC=C(C=C1)N2C(=O)C(=C(C=N2)Cl)Cl
Melting Point 164°C to 166°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Color Beige
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Formula Weight 241.08
Physical Form Crystalline Powder
Percent Purity ≥99.0%
Chemical Name or Material 1-Phenyl-4, 5-dichloro-6-pyridazone, 99+%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.