Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenylbiguanide hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
100mg
This item is not returnable. View return policy
55-57-2
C8H12ClN5
213.67
MFCD00035040
FHUDRDSKZQDCBC-UHFFFAOYSA-N
1-phenylbiguanide hydrochloride, phenylbiguanide hydrochloride, n-phenylimidodicarbonimidic diamide hydrochloride, phenyldiguanide hydrochloride, 1-phenylbiquanide hydrochloride, phenylbiguanide monohydrochloride, 1-phenylbiguanide monohydrochloride, n'-phenylbiguanide monohydrochloride, phenyl biguanide hydrochloride, imidodicarbonimidic diamide, n-phenyl-, monohydrochloride
5932
hydrogen N-(diaminomethylidene)-N''-phenylguanidine chloride
[H+].[Cl-].NC(N)=NC(N)=NC1=CC=CC=C1
This item is not returnable. View return policy
55-57-2
C8H12ClN5
213.67
MFCD00035040
FHUDRDSKZQDCBC-UHFFFAOYSA-N
1-phenylbiguanide hydrochloride, phenylbiguanide hydrochloride, n-phenylimidodicarbonimidic diamide hydrochloride, phenyldiguanide hydrochloride, 1-phenylbiquanide hydrochloride, phenylbiguanide monohydrochloride, 1-phenylbiguanide monohydrochloride, n'-phenylbiguanide monohydrochloride, phenyl biguanide hydrochloride, imidodicarbonimidic diamide, n-phenyl-, monohydrochloride
5932
hydrogen N-(diaminomethylidene)-N''-phenylguanidine chloride
[H+].[Cl-].NC(N)=NC(N)=NC1=CC=CC=C1

5-HT3 agonist

5-HT3 receptor agonist.
Quantity 100mg
Formula Weight 213.67
Percent Purity >98%
Chemical Name or Material 1-Phenylbiguanide hydrochloride

Recommended Storage : Store at Room Temperature