Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Phenylcyclopentanecarboxylic acid, 98%

Click to view available options
Quantity:
5g
25g
77-55-4
C12H14O2
190.242
MFCD00001372
RHPCYZLXNNRRMB-UHFFFAOYSA-N
1-phenylcyclopentanecarboxylic acid,1-phenyl-1-cyclopentanecarboxylic acid,cyclopentanecarboxylic acid, 1-phenyl,1-phenyl-cyclopentanecarboxylic acid,1-phenylcyclopentane carboxylic acid,cyclopentanecarboxylicacid, 1-phenyl,pubchem22923,acmc-209pao,phenylcyclopentanecarboxylic acid,phenylcyclopentane carboxylic acid
66167
1-phenylcyclopentane-1-carboxylic acid
C1CCC(C1)(C2=CC=CC=C2)C(=O)O
77-55-4
C12H14O2
190.242
MFCD00001372
RHPCYZLXNNRRMB-UHFFFAOYSA-N
1-phenylcyclopentanecarboxylic acid,1-phenyl-1-cyclopentanecarboxylic acid,cyclopentanecarboxylic acid, 1-phenyl,1-phenyl-cyclopentanecarboxylic acid,1-phenylcyclopentane carboxylic acid,cyclopentanecarboxylicacid, 1-phenyl,pubchem22923,acmc-209pao,phenylcyclopentanecarboxylic acid,phenylcyclopentane carboxylic acid
66167
1-phenylcyclopentane-1-carboxylic acid
C1CCC(C1)(C2=CC=CC=C2)C(=O)O
Melting Point 158°C to 161°C
Odor Odorless
Quantity 25g
Beilstein 1638653
Formula Weight 190.24
Percent Purity 98%
Chemical Name or Material 1-Phenylcyclopentanecarboxylic acid