Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyldodecane, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
123-01-3
C18H30
246.43
MFCD00008974
KWKXNDCHNDYVRT-UHFFFAOYSA-N
1-phenyldodecane,benzene, dodecyl,n-dodecylbenzene,laurylbenzene,phenyldodecane,marlican,dodecane, 1-phenyl,detergent alkylate,alkylate p 1,phenyldodecan
31237
dodecylbenzene
CCCCCCCCCCCCC1=CC=CC=C1
123-01-3
C18H30
246.43
MFCD00008974
KWKXNDCHNDYVRT-UHFFFAOYSA-N
1-phenyldodecane,benzene, dodecyl,n-dodecylbenzene,laurylbenzene,phenyldodecane,marlican,dodecane, 1-phenyl,detergent alkylate,alkylate p 1,phenyldodecan
31237
dodecylbenzene
CCCCCCCCCCCCC1=CC=CC=C1
Melting Point -7°C
Boiling Point 288°C to 300°C
Flash Point 124°C
Refractive Index 1.4810 to 1.483
Specific Gravity 0.85
Density 0.8500g/mL
Packaging Glass bottle
Quantity 250g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C6H5(CH2)11CH3
Solubility Information Solubility in water: immiscible with water
Formula Weight 246.43
Chemical Name or Material 1-Phenyldodecane
Show More Show Less