Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Phenylnaphthalene, 97%

Click to view available options
Quantity:
5g
25g
100g
605-02-7
C16H12
204.272
MFCD00003983
IYDMICQAKLQHLA-UHFFFAOYSA-N
naphthalene, 1-phenyl,1-phenyl-naphthalene,unii-10nxc4g45q,phenylnaphthalene,phenyl-napthalene,p-4-naphthyl benzene,acmc-1bfcg,1-phenylnaphthalene,naphthalene, 1-phenyl-8ci 9ci,10 microg/ml in acetonitrile
11795
1-phenylnaphthalene
C1=CC=C(C=C1)C2=CC=CC3=CC=CC=C32
605-02-7
C16H12
204.272
MFCD00003983
IYDMICQAKLQHLA-UHFFFAOYSA-N
naphthalene, 1-phenyl,1-phenyl-naphthalene,unii-10nxc4g45q,phenylnaphthalene,phenyl-napthalene,p-4-naphthyl benzene,acmc-1bfcg,1-phenylnaphthalene,naphthalene, 1-phenyl-8ci 9ci,10 microg/ml in acetonitrile
11795
1-phenylnaphthalene
C1=CC=C(C=C1)C2=CC=CC3=CC=CC=C32
Boiling Point 320°C
Quantity 100g
Flash Point >110°C (230°F)
Beilstein 1906885
Formula Weight 204.27
Percent Purity 97%
Chemical Name or Material 1-Phenylnaphthalene