Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenylpiperazine, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
92-54-6
C10H14N2
162.24
MFCD00005957
YZTJYBJCZXZGCT-UHFFFAOYSA-N
phenylpiperazine,n-phenylpiperazine,piperazine, 1-phenyl,1-phenyl-piperazine,n-phenyldiethylenediamine,1-fenylpiperazin czech,unii-j9225cbi7d,n-phenyl piperazine,1-phenyl piperazine,ccris 4334
7096
1-phenylpiperazine
C1CN(CCN1)C2=CC=CC=C2
92-54-6
C10H14N2
162.24
MFCD00005957
YZTJYBJCZXZGCT-UHFFFAOYSA-N
phenylpiperazine,n-phenylpiperazine,piperazine, 1-phenyl,1-phenyl-piperazine,n-phenyldiethylenediamine,1-fenylpiperazin czech,unii-j9225cbi7d,n-phenyl piperazine,1-phenyl piperazine,ccris 4334
7096
1-phenylpiperazine
C1CN(CCN1)C2=CC=CC=C2
Specific Gravity 1.062
Color Colorless to Yellow
Flash Point 140°C
Refractive Index 1.5865 to 1.5885
Infrared Spectrum Authentic
Quantity 25g
Melting Point 18.8°C
Packaging Glass bottle
Boiling Point 286.0°C
Water 0.5% max. (K.F.)
Density 1.0620g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
UN Number 2927
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 162.24
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 1-Phenylpiperazine
Show More Show Less

GHS H Statement:
Fatal in contact with skin.
Toxic if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger