Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenylpyrrolidine, 98+%, Thermo Scientific Chemicals

Catalog Number p-7058299
Click to view available options
Quantity:
5 g
25 g
4096-21-3
C10H13N
147.22
MFCD00015897
VDQQJMHXZCMNMU-UHFFFAOYSA-N
1-phenyl-pyrrolidine, n-phenylpyrrolidine, phenylpyrrolidine, pyrrolidine, 1-phenyl, 1-phenyl-pyrrolidin-3-ol, 1-phenylpyrolidine, n-phenyl pyrrolidine, pubchem9192, acmc-1cttc
77726
1-phenylpyrrolidine
C1CCN(C1)C1=CC=CC=C1
4096-21-3
C10H13N
147.22
MFCD00015897
VDQQJMHXZCMNMU-UHFFFAOYSA-N
1-phenyl-pyrrolidine, n-phenylpyrrolidine, phenylpyrrolidine, pyrrolidine, 1-phenyl, 1-phenyl-pyrrolidin-3-ol, 1-phenylpyrolidine, n-phenyl pyrrolidine, pubchem9192, acmc-1cttc
77726
1-phenylpyrrolidine
C1CCN(C1)C1=CC=CC=C1

1-Phenylpyrrolidine are used as flooding intestinal coccidiosis, flooding Xiao NET intermediate.It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenylpyrrolidine are used as flooding intestinal coccidiosis, flooding Xiao NET intermediate.It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents. Store cold 2-8°C.
TRUSTED_SUSTAINABILITY
Boiling Point 133°C to 134°C (19 mmHg)
Refractive Index 1.582
Quantity 5 g
Beilstein 117030
Solubility Information Not miscible in water.
Formula Weight 147.22
Percent Purity ≥98%
Chemical Name or Material 1-Phenylpyrrolidine
Hazard Category H302+H312
Hazard Statement GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
EINECSNumber 223-849-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit