Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Phenylsulfonyl)indole, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
40899-71-6
C14H11NO2S
257.307
MFCD00134318
VDWLCYCWLIKWBV-UHFFFAOYSA-N
1-phenylsulfonyl-1h-indole,1-phenylsulfonyl indole,1-benzenesulfonyl indole,1-benzenesulfonyl-1h-indole,n-phenylsulfonylindole,1-phenylsulphonyl indole,1-phenylsulfonyl-indole,chembl82224
315017
1-(benzenesulfonyl)indole
C1=CC=C(C=C1)S(=O)(=O)N2C=CC3=CC=CC=C32
40899-71-6
C14H11NO2S
257.307
MFCD00134318
VDWLCYCWLIKWBV-UHFFFAOYSA-N
1-phenylsulfonyl-1h-indole,1-phenylsulfonyl indole,1-benzenesulfonyl indole,1-benzenesulfonyl-1h-indole,n-phenylsulfonylindole,1-phenylsulphonyl indole,1-phenylsulfonyl-indole,chembl82224
315017
1-(benzenesulfonyl)indole
C1=CC=C(C=C1)S(=O)(=O)N2C=CC3=CC=CC=C32
Quantity 1g
Color Pink to White
Melting Point 73°C to 79°C
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 257.31
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 1-(Phenylsulfonyl)indole
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.