Alfa Aesar™ 1-Phenylsulfonylpyrazole, 95%

Manufacturer: Alfa Aesar™ H3291403

Quantity 1g

 View more versions of this product

Catalog No. AAH3291403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 108128-27-4
    Molecular Formula C9H8N2O2S
    Formula Weight 208.24
    MDL Number MFCD00514543
    Synonym 1-Benzenesulfonylpyrazole
    Chemical Name or Material 1-Phenylsulfonylpyrazole

    Specifications

    Formula Weight 208.24
    Molecular Formula C9H8N2O2S
    MDL Number MFCD00514543
    CAS 108128-27-4
    Synonym 1-Benzenesulfonylpyrazole
    Quantity 1g
    Chemical Name or Material 1-Phenylsulfonylpyrazole
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit