Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Piperazinecarboxaldehyde, 90%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
7755-92-2
C5H10N2O
114.15
MSSDTZLYNMFTKN-UHFFFAOYSA-N
1-formylpiperazine,1-piperazinecarboxaldehyde,1-piperazinecarbaldehyde,formylpiperazine,n-formylpiperazine,1-formyl piperazine,piperazinecarbaldehyde,l-formylpiperazine,4-formylpiperazine,n-formyl piperazine
82191
piperazine-1-carbaldehyde
C1CN(CCN1)C=O
7755-92-2
C5H10N2O
114.15
MSSDTZLYNMFTKN-UHFFFAOYSA-N
1-formylpiperazine,1-piperazinecarboxaldehyde,1-piperazinecarbaldehyde,formylpiperazine,n-formylpiperazine,1-formyl piperazine,piperazinecarbaldehyde,l-formylpiperazine,4-formylpiperazine,n-formyl piperazine
82191
piperazine-1-carbaldehyde
C1CN(CCN1)C=O
Boiling Point 105.0°C to 107.0°C (1.0 mmHg)
Specific Gravity 1.1
Refractive Index 1.5097 to 1.5117
Infrared Spectrum Authentic
Packaging Glass bottle
Flash Point 102°C
Density 1.1000g/mL
Quantity 25g
Color Colorless to Yellow
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 88% min. (GC)
Formula Weight 114.15
Physical Form Liquid
Percent Purity 90%
Chemical Name or Material 1-Piperazinecarboxaldehyde
Show More Show Less

GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water

GHS Signal Word: Danger

EINECSNumber : 231-813-