Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Piperidinecarbonitrile, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
50g
Packaging:
Glass bottle
1530-87-6
C6H10N2
110.16
MFCD00006477
NVPICXQHSYQKGM-UHFFFAOYSA-N
1-piperidinecarbonitrile,1-cyanopiperidine,piperidinocarbonitrile,n-cyanopiperidine,pentamethylenecyanamide,cyanopiperidine,cyano-piperidine,piperidine nitrile,piperidinecarbonitrile,1-cyanopiperidine #
73736
piperidine-1-carbonitrile
C1CCN(CC1)C#N
1530-87-6
C6H10N2
110.16
MFCD00006477
NVPICXQHSYQKGM-UHFFFAOYSA-N
1-piperidinecarbonitrile,1-cyanopiperidine,piperidinocarbonitrile,n-cyanopiperidine,pentamethylenecyanamide,cyanopiperidine,cyano-piperidine,piperidine nitrile,piperidinecarbonitrile,1-cyanopiperidine #
73736
piperidine-1-carbonitrile
C1CCN(CC1)C#N
Refractive Index 1.4690 to 1.4710 (20°C, 589nm)
Color Colorless
Density 0.9500g/mL
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 20, 56
Formula Weight 110.16
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 1-Piperidinecarbonitrile, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 216-233-8

TSCA : TSCA